Learn More
Medchemexpress LLC Fingolimod phosphate | 402615-91-2 | C19H34NO5P | 5 MG
SDP
Catalog No. 5000376032
Encompass_Preferred
Fingolimod phosphate is an orally active sphingosine 1-phosphate (S1P) receptor agonist. It promotes the neuroprotective effects of microglia and is used for the research of multiple sclerosis and neurologic diseases. In vitro, it prevents lymphocytes from moving out of lymphoid organs and inhibits autoreactive lymphocytes from infiltrating the central nervous system.
  • Orally active sphingosine 1-phosphate (S1P) receptor agonist
  • Promotes neuroprotective effects of microglia
  • Used for research of multiple sclerosis and neurologic diseases
  • Prevents lymphocytes from moving out of lymphoid organs
  • Inhibits autoreactive lymphocytes from infiltrating the central nervous system
  • Downregulates activated microglial production of pro-inflammatory cytokines
  • Upregulates microglial production of brain-derived neurotrophic factor and glial cell-derived neurotrophic factor
Catalog No. 50-003-76032 Supplier Medchemexpress LLC Supplier No. HY153815MG
Fingolimod phosphate is an orally active sphingosine 1-phosphate (S1P) receptor agonist. It promotes the neuroprotective effects of microglia and is used for the research of multiple sclerosis and neurologic diseases. In vitro, it prevents lymphocytes from moving out of lymphoid organs and inhibits autoreactive lymphocytes from infiltrating the central nervous system.
  • Orally active sphingosine 1-phosphate (S1P) receptor agonist
  • Promotes neuroprotective effects of microglia
  • Used for research of multiple sclerosis and neurologic diseases
  • Prevents lymphocytes from moving out of lymphoid organs
  • Inhibits autoreactive lymphocytes from infiltrating the central nervous system
  • Downregulates activated microglial production of pro-inflammatory cytokines
  • Upregulates microglial production of brain-derived neurotrophic factor and glial cell-derived neurotrophic factor