Learn More
MSC SPI Surface Finish Comparator, 0.1-3.2 Ra μm, Nickel, 6 Specimens
Catalog No. 5000536094
Encompass
This surface finish comparator is designed to provide a visual and tactile comparison of surface finishes. It includes six specimens that cover 0.1 to 3.2 Ra μm and 4 to 125 AA microinch surface finishes, making it suitable for various quality control and inspection tasks. The comparator comes with instructions and a protective case.
  • Includes six specimens for surface comparison
  • Covers 0.1 to 3.2 Ra μm and 4 to 125 AA microinch surface finishes
  • Constructed from nickel material
  • Overall dimensions of 2.5 in length by 5 in width
  • Supplied with a protective case and instructions
  • Complies with SAE, ANSI-B-46.1-1978, and Military Specifications 45662 standards
  • Machining process: belt sanding
Catalog No. 50-005-36094 Supplier MSC Supplier No. 06558266
This surface finish comparator is designed to provide a visual and tactile comparison of surface finishes. It includes six specimens that cover 0.1 to 3.2 Ra μm and 4 to 125 AA microinch surface finishes, making it suitable for various quality control and inspection tasks. The comparator comes with instructions and a protective case.
  • Includes six specimens for surface comparison
  • Covers 0.1 to 3.2 Ra μm and 4 to 125 AA microinch surface finishes
  • Constructed from nickel material
  • Overall dimensions of 2.5 in length by 5 in width
  • Supplied with a protective case and instructions
  • Complies with SAE, ANSI-B-46.1-1978, and Military Specifications 45662 standards
  • Machining process: belt sanding