Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
GSA_VAGREENER_CHOICE

Catalog No. 05719510 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.2 to 1000 μL
1 to 1000 μL
10 to 10,000 μL
2 to 1000 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-719-510 1 to 1000 μL
05-719-511 0.2 to 1000 μL
05-719-512 10 to 10,000 μL
05-719-621 2 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05-719-510 Supplier Thermo Scientific™ Supplier No. 4700870
  • $2,390.00
    $1,714.65 / Each of 1

    Save $675.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enjoy the convenience of having a variety of Finnpipette™ F2 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab

Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.

Features:

  • AVG-volume gearing mechanism for accuracy and precision
  • Fully autoclavable for protection
  • Soft-touch tip ejector for light tip ejection
  • Large display for better vision ergonomics
  • Very light pipetting forces

Kit 1 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 1 to 1000 μL:
    • Finnpipette F2
    • 1-10 μL
    • 10-100 μL
    • 100-1000 μL
  • Flex tips: 10, 1 x 96; 200, 1 x 96; 1000, 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000 μL:
    • Finnpipette F2
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Flex tips: 10, 12 x 96; 200, 2 x 96; 1000, 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 10 to 10,000 μL:
    • Finnpipette F2
    • 10-100 μL 100-1000 μL
    • 1000-10,000 μL
  • Flex tips: 200, 1 x 96; 1000, 1 x 96; FT 10 mL, 1 x 24 F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 20 to 1000 μL:
    • Finnpipette F2
    • 2-20 μL 20-200 μL
    • 100-1000 μL 
  • Flex tips: (2) 200μL, 10 x 96; 100-1000 μL, 10 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains all the information of pipetting from choosing the right type to detailed decontamination information.

Warranty: 5 years with web registration.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips Flex Tips: 10, 200, 1000 μL
Description Finnpipette F2 GLP Kit 1: 1-10μL micro, 10-100μL and 100-1000μL
Autoclavable Autoclavable
Color Assorted
Model F2
Number of Channels 1
Product Line Finnpipette F2
Type GLP Kit
Volume (Metric) 1 to 1000 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.