Learn More
Thermo Scientific™ Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits

Shop All Thermo Scientific Products


Description
Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.
Features:
- AVG-volume gearing mechanism for accuracy and precision
- Fully autoclavable for protection
- Soft-touch tip ejector for light tip ejection
- Large display for better vision ergonomics
- Very light pipetting forces
Kit 1 includes:
- Three Thermo Scientific Finnpipette Pipetters from 1 to 1000 μL:
- Finnpipette F2
- 1-10 μL
- 10-100 μL
- 100-1000 μL
- Flex tips: 10, 1 x 96; 200, 1 x 96; 1000, 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 2 includes:
- Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000 μL:
- Finnpipette F2
- 0.2-2 μL
- 2-20 μL univ
- 20-200 μL
- 100-1000 μL
- Flex tips: 10, 12 x 96; 200, 2 x 96; 1000, 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 3 includes:
- Three Thermo Scientific Finnpipette Pipetters from 10 to 10,000 μL:
- Finnpipette F2
- 10-100 μL 100-1000 μL
- 1000-10,000 μL
- Flex tips: 200, 1 x 96; 1000, 1 x 96; FT 10 mL, 1 x 24 F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 4 includes:
- Three Thermo Scientific Finnpipette Pipetters from 20 to 1000 μL:
- Finnpipette F2
- 2-20 μL 20-200 μL
- 100-1000 μL
- Flex tips: (2) 200 μL, 10 x 96; 100-1000 μL, 10 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Ordering Information:
The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains all the information of pipetting from choosing the right type to detailed decontamination information.
Warranty: 5 years with web registration.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Compatible Tips | Flex Tips: 200, 1000 μL, 10 mL |
| Description | Finnpipette F2 GLP Kit 3: 10-100μL, 100-1000μL and 1-10mL |
| Autoclavable | Autoclavable |
| Color | Assorted |
| Model | F2 |
| Number of Channels | 1 |
| Product Line | Finnpipette F2 |
| Type | GLP Kit |
| Volume (Metric) | 10 to 10,000 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.