Learn More
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipette PROMO
Catalog No. 14386316TI Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
2 to 20 μL
5 to 50 μL
Color:
Turquoise
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14-386-316TI 2 to 20 μL Turquoise
14-386-294TI 5 to 50 μL Turquoise
14-386-295TI 5 to 50 μL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-386-316TI Supplier Thermo Scientific™ Supplier No. P4642050
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of these fully autoclavable pipettes.
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling. They feature a wide selection of expressly designed tips for optimal performance, precision and accuracy.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use:
  • Rugged and Durable: Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
  • Fully autoclavable: Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
  • Maintenance is Easy: Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
  • Super Blow-out: 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50-μL models and below.
  • Wide Selection of Finntip Pipette Tips: Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
  • CE marked: The Finnpipette F2 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
  • 5 Year Warranty: With convenient online registration, and may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Finntip 50 Micro, Filter 50 Micro
Description Finnpipette F2
Autoclavable Autoclavable
Color Turquoise
Number of Channels 1
Product Line Finnpipette
Type Variable Volume Pipette
Volume (Metric) 2 to 20 μL
Increments 0.02 μL
Quantity 1 Ea.