Learn More
Description
Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ R&D Systems GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ R&D Systems™ pipetters, appropriate stands and sample tips.
R&D Systems GLP Kit 1 includes:
- Finnpipette R&D Systems 10-100μL
- Finnpipette R&D Systems 100-1000μL
- Two Finnpipette R&D Systems stands
- One rack of Finntip Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Finnpipette R&D Systems brochure
R&D Systems GLP Kit 2 includes:
- Finnpipette R&D Systems 100-1000μL
- Finnpipette R&D Systems 0.5-5mL
- Two Finnpipette R&D Systems stands
- One rack of Finntip 5ml and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Finnpipette R&D Systems brochure
- Pipetting Guide
R&D Systems GLP Kit 3 includes:
- R&D Systems pipette, 1-10μL, micro
- R&D Systems pipette, 10-100μL
- R&D Systems pipette, 100-1000μL
- Three Finnpipette R&D Systems pipette stands
- One rack of Flex 10, Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- R&D Systems brochure
R&D Systems GLP Kit 4 includes:
- R&D Systems pipette, 10-100μL
- R&D Systems pipette, 100-1000μL
- R&D Systems pipette, 1-10mL
- Three Finnpipette R&D Systems pipette stands
- One rack of Flex 200, Flex 1000 and Finntip 10ml
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- R&D Systems brochure
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Assorted |
| Model | Novus |
| Number of Channels | 1 |
| Product Line | Novus |
| Type | GLP Kit |
| Volume (Metric) | 100 to 5000 μL |
| Includes | Novus 1000 and 5000 μL single channel pipettes, two Novus stands, two sample Finntip racks |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

