Learn More
Pipette Supplies, Inc. FINNPIPETTE ORING 10UL
SDP
Supplier:  Pipette Supplies, Inc. TS 1030060
Encompass
O-ring for:E1 ClipTip Single Channel 12.5μL (piston o-ring), 125μL & 300μL (metal tip fitting upper o-ring)E1 ClipTip Multichannel, 125μL, 300μL, 850μL, 1250μL (metal tip fitting upper o-ring)Finnpipette BioControl Multichannel 10μLFinnpipette Campus Single Channel 10μLFinnpipette Colour Single Channel 10μLFinnpipette Digital Single Channel 10μL, 10μL microFinnpipette Digital Multichannel 10μL microFinnpipette F1 Single Channel 10μL, 10μL microFinnpipette F1 Multichannel, 16 Channel 10μLFinnpipette F1 ClipTip Single Channel 10μl (piston o-ring), 50μL, 100μL, 200μL & 300μL (metal tip fitting upper o-ring)Finnpipettte F1 ClipTip Fixed Volume 1μl, 5μl, 10μl, 20μl, 25μl, 50μl, 100μl, 200μl, 250μlFinnpipette F1 ClipTip Multichannel, All Volumes (metal tip fitting upper o-ring)Finnpipette F2 Single Channel 10μL, 10μL microFinnpipette F3 Single Channel 10μL, 10μL micro
Catalog No. 50-131-7656