Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips
GSA_VAGREENER_CHOICE

Catalog No. 21377162 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
21-377-162 Non-sterile
21-377-163 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-162 Supplier Thermo Scientific™ Supplier No. 9420300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Combine these Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips with your pipette.

Combine these Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips with your pipette. Choose sterile or nonsterile.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description For 25 or 50mL Finnitip Stepper tips
For Use With FinnTip Stepper Tip (25, 50 mL)
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.