Learn More
Sigma Aldrich Fine Chemicals Biosciences finnpipetteR f2 variable v1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6783411EA
Encompass
finnpipette f2 has a light pipetting action is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the finnpipette f2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision Big display with white numbers on black background for increased visibility Double action pipetting button with very light and smooth pipetting actionfully autoclavableSuper blow-out piston with volumes of 50mul and below
Catalog No. 50-290-5552