Learn More
Sigma Aldrich Fine Chemicals Biosciences finnpipetteR pipette tips
Supplier: Sigma Aldrich Fine Chemicals Biosciences P704910EA
Each tray contains 8AxA12A(96) tips. When used with multi-channel pipettes spacing between tips in tray permits mounting up to 12 tips at once.