Learn More
Thermo Scientific™ Finntip™ Biocon Pipette Tips
Catalog No. 21377358
Change view
Click to view available options
Volume (Metric):
10 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-377-358 10 μL
21-377-360 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-358 Supplier Thermo Scientific™ Supplier No. 94053100
Perform PCR and other purity demanding applications with these biocon pipette tips, certified free from endotoxin, DNA, DNase and RNase.
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Features:
  • Individually packed
  • Three models in volume range 0.2-1000 μL
  • Presterilized with gamma radiation
  • Includes certificate of quality
Recommended for: PCR applications, molecular biology
TRUSTED_SUSTAINABILITY
Specifications
Description Finntip BioCon 10, Non-Filtered, Sterile,
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Model Finntip BioCon 10
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
For Use With (Equipment) Thermo Scientific FinnTip BioCon 10
Product Line FinnTip Biocon