Learn More
Thermo Scientific™ Finntip™ Biocon Pipette Tips
Description
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
- Individually packed
- Three models in volume range 0.2-1000 μL
- Presterilized with gamma radiation
- Includes certificate of quality
Recommended for: PCR applications, molecular biology
Specifications
Specifications
| Description | Finntip BioCon 1000, Non-Filtered, Sterile, |
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Model | Finntip BioCon 1000 |
| Certifications/Compliance | ISO 9001, ISO 14001, ISO 13485 |
| For Use With (Equipment) | Thermo Scientific FinnTip BioCon 1000 |
| Product Line | FinnTip Biocon |