Learn More
Thermo Scientific™ Finntip™ Specific Pipette Tips
Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Supplier:  Thermo Scientific™ 9402151
Catalog No. 21-377-52
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Description
Description
Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2μL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk, and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging
Note:
Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified)
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions