Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finntip™ Universal Pipette Tips

Catalog No. 21377144 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Finntip 250 Universal, Non-Filtered, Non-sterile, Box
Volume (Metric):
250 μL
Format:
Box
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Volume (Metric) Format Sterility
21-377-144 Finntip 250 Universal, Non-Filtered, Non-sterile, Box 250 μL Box Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-144 Supplier Thermo Scientific™ Supplier No. 9400250

missing translation for 'validMainframeMsg'

Ensure accurate, precise pipetting with these universal pipette tips, designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Ensure accurate, precise pipetting with Thermo Scientific™ Finntip™ Universal Pipette Tips, manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips to prevent liquid retention.

Features:

  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2μL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging

Note:

Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified).


Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warrantied from the date we ship the product and for ninety (90) days thereafter.

TRUSTED_SUSTAINABILITY

Specifications

Description Finntip 250 Universal, Non-Filtered, Non-sterile, Box
Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Model Finntip 250 Universal
Length (English) 2 in.
Length (Metric) 5.2cm
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
Format Box
Tip Style Universal Fit
Packaging 500 tips
For Use With (Equipment) Thermo Scientific Finnpipette 250
Product Line FinnTip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.