Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips, 100-1000 μL, Tray
GREENER_CHOICE

Catalog No. 21377602 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
100 to 1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-377-602 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-602 Supplier Thermo Scientific™ Supplier No. 9405160
  • $287.00
    $225.65 / Case of 960

    Save $61.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips, ideal for pipetting macromolecules.

  • Two different models cover volume range of 10-1000 μL
  • Wide orifice tip; inside diameter of orifice is 1.1mm/1.8mm
  • Color code: White
  • Length: 5.3cm/7.2cm
TRUSTED_SUSTAINABILITY

Specifications

Description Finntip 1000 Wide, Non-Filtered, Non-sterile, Rack
Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Model Finntip 1000 Wide
Length (English) 2.8 in.
Length (Metric) 7.2cm
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
Format Rack
Packaging 10 x 96/Rack
For Use With (Equipment) Thermo Scientific Finnpipette
Product Line FinnTip Wide Orifice
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.