Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips
GREENER_CHOICE

Catalog No. 21377119 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Format
21-377-119 250 μL Sterile Rack
21-377-117B 250 μL Non-sterile Bagged
21-377-118B 1000 μL Non-sterile Bagged
21-377-603 1000 μL Sterile Rack
21-377-117C 250 μL Non-sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 21-377-119 Supplier Thermo Scientific™ Supplier No. 9405123
  • $320.00
    $224.65 / Case of 960

    Save $95.35

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate cell fragmentation caused by mechanical shearing with these wide orifice pipette tips, ideal for pipetting macromolecules.

Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips, ideal for pipetting macromolecules.

  • Two different models cover volume range of 10-1000 μL
  • Wide orifice tip; inside diameter of orifice is 1.1mm/1.8mm
  • Color code: White
  • Length: 5.3cm/7.2cm
TRUSTED_SUSTAINABILITY

Specifications

Description Finntip 250 Wide Sterile, Non-Filtered, Sterile, Rack
Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Sterile
Model Finntip 250 Wide Sterile
Length (English) 2 in.
Length (Metric) 5.3 cm
Format Rack
Tip Style Wide Orifice
Color Code White
For Use With (Equipment) Thermo Scientific Finnpipette
Product Line FinnTip Wide Orifice
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.