Learn More
Sigma Aldrich Fine Chemicals Biosciences finntipR flex pipette tips
Catalog No. 502901511
Encompass
finntip flex pipette tips are easy-to-use soft flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface which prevents liquid retention and achieves better pipetting accuracy.
Catalog No. 50-290-1511 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z677876960EA
finntip flex pipette tips are easy-to-use soft flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface which prevents liquid retention and achieves better pipetting accuracy.