Learn More
Aviva Systems Biology Corp FIP1L1 Antibody - C-terminal region (ARP41031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FIP1L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FIP1L1. Alternate names: DKFZp586K0717, FLJ33619, Rhe, hFip1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53264