Learn More
Medchemexpress LLC Fipexide hydrochloride | 34161-23-4 | 25 MG
SDP
Catalog No. 5000413928
Encompass_Preferred
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and positively affects cognitive performance through dopaminergic neurotransmission. This product is intended for research and analytical applications, particularly in senile dementia research.
  • Orally active nootropic agent
  • Reduces striatal adenylate cyclase activity
  • Positive effect on cognitive performance by dopaminergic neurotransmission
  • Used for senile dementia research
  • Acts as a chemical inducer in callus formation, shoot regeneration and Agrobacterium infection
  • Intended for research and analytical applications
Catalog No. 50-004-13928 Supplier Medchemexpress LLC Supplier No. 25MGHYB1124A
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and positively affects cognitive performance through dopaminergic neurotransmission. This product is intended for research and analytical applications, particularly in senile dementia research.
  • Orally active nootropic agent
  • Reduces striatal adenylate cyclase activity
  • Positive effect on cognitive performance by dopaminergic neurotransmission
  • Used for senile dementia research
  • Acts as a chemical inducer in callus formation, shoot regeneration and Agrobacterium infection
  • Intended for research and analytical applications