Learn More
Medchemexpress LLC Fipexide hydrochloride | 34161-23-4 | 99.88% | 1 ML
SDP
Catalog No. 5000413488
Encompass_Preferred
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and has a positive effect on cognitive performance through dopaminergic neurotransmission. It is used for senile dementia research and acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Parachloro-phenossiacetic acid derivative.
  • Orally active nootropic agent.
  • Reduces striatal adenylate cyclase activity.
  • Positive effect on cognitive performance.
  • Used for senile dementia research.
  • Chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
Catalog No. 50-004-13488 Supplier Medchemexpress LLC Supplier No. HYB1124A10MM/1ML
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and has a positive effect on cognitive performance through dopaminergic neurotransmission. It is used for senile dementia research and acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Parachloro-phenossiacetic acid derivative.
  • Orally active nootropic agent.
  • Reduces striatal adenylate cyclase activity.
  • Positive effect on cognitive performance.
  • Used for senile dementia research.
  • Chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.