Learn More
Medchemexpress LLC Fipexide hydrochloride | 34161-23-4 | 99.9% | 50 MG
SDP
Catalog No. 5000413072
Encompass_Preferred
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and positively affects cognitive performance through dopaminergic neurotransmission. Fipexide hydrochloride is used for senile dementia research. It also acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Reduces striatal adenylate cyclase activity.
  • Positively affects cognitive performance.
  • Used in senile dementia research.
  • Acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Abolishes memory deficits produced by kindling.
Catalog No. 50-004-13072 Supplier Medchemexpress LLC Supplier No. HYB1124A50MG
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and positively affects cognitive performance through dopaminergic neurotransmission. Fipexide hydrochloride is used for senile dementia research. It also acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Reduces striatal adenylate cyclase activity.
  • Positively affects cognitive performance.
  • Used in senile dementia research.
  • Acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Abolishes memory deficits produced by kindling.