Learn More
Medchemexpress LLC Fipexide hydrochloride | 34161-23-4 | MFCD00133323 | 99.8% | 100 MG
SDP
Catalog No. 5000411325
Encompass_Preferred
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and positively affects cognitive performance through dopaminergic neurotransmission. It is used in senile dementia research and acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Orally active nootropic agent.
  • Reduces striatal adenylate cyclase activity.
  • Positive effect on cognitive performance by dopaminergic neurotransmission.
  • Used for senile dementia research.
  • Acts as a chemical inducer in callus formation, shoot regeneration and Agrobacterium infection.
Catalog No. 50-004-11325 Supplier Medchemexpress LLC Supplier No. HYB1124A100MG
Fipexide hydrochloride is a parachloro-phenossiacetic acid derivative and an orally active nootropic agent. It reduces striatal adenylate cyclase activity and positively affects cognitive performance through dopaminergic neurotransmission. It is used in senile dementia research and acts as a chemical inducer in callus formation, shoot regeneration, and Agrobacterium infection.
  • Orally active nootropic agent.
  • Reduces striatal adenylate cyclase activity.
  • Positive effect on cognitive performance by dopaminergic neurotransmission.
  • Used for senile dementia research.
  • Acts as a chemical inducer in callus formation, shoot regeneration and Agrobacterium infection.