Learn More
Enzo Life Sciences Fipronil (100mg). CAS: 120068-37-3
SDP
Catalog No. 502010413
Encompass_Preferred
GABA-gated chloride channel blocker. Blocks GABA responses on Xenopus oocytes. Blocks GABA-gated chloride ion channel in Drosophila. Purity: ≥98%. Long Term Storage: Ambient.
Catalog No. 50-201-0413 Supplier Enzo Life Sciences Supplier No. BMLCM1020100
GABA-gated chloride channel blocker. Blocks GABA responses on Xenopus oocytes. Blocks GABA-gated chloride ion channel in Drosophila. Purity: ≥98%. Long Term Storage: Ambient.