Learn More
Cayman Chemical Fipronil-13C2,15N2, Purity: ≥95%, 1 mg
SDP
Catalog No. 502915927
Encompass_Preferred
An internal standard for the quantification of fipronil by GC- or LC-MS
Catalog No. 50-291-5927 Supplier Cayman Chemical Supplier No. 395871 MG
An internal standard for the quantification of fipronil by GC- or LC-MS