Learn More
AdipoGen Fipronil
SDP
Catalog No. 502562252
Encompass_Preferred
Chemical. CAS 120068-37-3. Formula C12H4Cl2F6N4OS. MW 437.2. Alzheimer inducing agent Alzheimerogen. Potent GABA receptor alpha-4 subunit and beta-3 subunit inhibitor. Broad-range insecticide. Blocks GABA A-gated chloride channels and glutamate-activated chloride channels GluCls in the central nervous system, resulting in overexcitation of the insects nervous system and muscles. Induces oxidative stress in mouse kidney and brain. Increases CYP1A1, CYP2B6, CYP3A4 and CYP3A5 transcript, protein and enzyme activity levels.
Catalog No. 50-256-2252 Supplier AdipoGen Supplier No. AGCR13648G001
Chemical. CAS 120068-37-3. Formula C12H4Cl2F6N4OS. MW 437.2. Alzheimer inducing agent Alzheimerogen. Potent GABA receptor alpha-4 subunit and beta-3 subunit inhibitor. Broad-range insecticide. Blocks GABA A-gated chloride channels and glutamate-activated chloride channels GluCls in the central nervous system, resulting in overexcitation of the insects nervous system and muscles. Induces oxidative stress in mouse kidney and brain. Increases CYP1A1, CYP2B6, CYP3A4 and CYP3A5 transcript, protein and enzyme activity levels.