Learn More
Medchemexpress LLC Fipronil sulfone | 120068-36-2 | MFCD01938707 | 99.8% | 453.15 g/mol | C12H4Cl2F6N4O2S | 10MG
SDP
Catalog No. 5000236359
Encompass_Preferred
Fipronil sulfone is the major metabolite of the insecticide fipronil and acts as a GABA receptor inhibitor. Supplied as a high-purity analytical/research standard, it is suitable for in vitro pharmacology, metabolic and environmental analysis, and use as a reference material.
  • High purity (99.8%).
  • Suitable for pharmacology and in vitro studies.
  • Reported GABA receptor IC50 ≈ 175 nM.
  • Molecular weight 453.15 g/mol; formula C12H4Cl2F6N4O2S.
  • Available in small pack sizes for analytical applications.
Catalog No. 50-002-36359 Supplier Medchemexpress LLC Supplier No. HY12529610MG
Fipronil sulfone is the major metabolite of the insecticide fipronil and acts as a GABA receptor inhibitor. Supplied as a high-purity analytical/research standard, it is suitable for in vitro pharmacology, metabolic and environmental analysis, and use as a reference material.
  • High purity (99.8%).
  • Suitable for pharmacology and in vitro studies.
  • Reported GABA receptor IC50 ≈ 175 nM.
  • Molecular weight 453.15 g/mol; formula C12H4Cl2F6N4O2S.
  • Available in small pack sizes for analytical applications.