Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fipronil sulfone | 120068-36-2 | MFCD01938707 | 99.8% | 453.15 g·mol⁻¹ | C12H4Cl2F6N4O2S | 25MG
SDP

Catalog No. 5000237234
Encompass_Preferred

Fipronil sulfone is the primary oxidative metabolite of fipronil, supplied as a research-grade small molecule for biochemical, pharmacological, and analytical studies. It acts as a GABA receptor inhibitor and is used as an analytical reference, in metabolism investigations, receptor binding assays, and environmental monitoring.

  • High chemical purity (99.8%).
  • White to off-white solid, stable under recommended storage conditions.
  • Molecular weight 453.15 g·mol⁻¹ and formula C12H4Cl2F6N4O2S.
  • Soluble in DMSO at 125 mg/mL; ultrasonic assistance may be required.
  • Available in small milligram pack sizes suitable for laboratory research.

Catalog No. 50-002-37234 Supplier Medchemexpress LLC Supplier No. HY12529625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fipronil sulfone is the primary oxidative metabolite of fipronil, supplied as a research-grade small molecule for biochemical, pharmacological, and analytical studies. It acts as a GABA receptor inhibitor and is used as an analytical reference, in metabolism investigations, receptor binding assays, and environmental monitoring.

  • High chemical purity (99.8%).
  • White to off-white solid, stable under recommended storage conditions.
  • Molecular weight 453.15 g·mol⁻¹ and formula C12H4Cl2F6N4O2S.
  • Soluble in DMSO at 125 mg/mL; ultrasonic assistance may be required.
  • Available in small milligram pack sizes suitable for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.