Learn More
Medchemexpress LLC Fipronil sulfone | 120068-36-2 | MFCD01938707 | 99.8% | 453.15 g/mol | C12H4Cl2F6N4O2S | 5MG
SDP
Catalog No. 5000238347
Encompass_Preferred
Fipronil sulfone is the sulfone metabolite of fipronil supplied as an analytical standard for research and method development. It is used as a reference material in studies of metabolism, environmental monitoring, and receptor-binding assays, and is reported to selectively inhibit the GABA receptor (IC50 ~175 nM).
  • High-purity analytical standard (reported 99.8%).
  • Molecular formula C12H4Cl2F6N4O2S and molecular weight 453.15 g/mol.
  • Suitable for method validation and quantification of fipronil metabolites.
  • Demonstrated GABA receptor activity useful for biochemical studies.
  • Available in small pack sizes appropriate for analytical workflows.
Catalog No. 50-002-38347 Supplier Medchemexpress LLC Supplier No. HY1252965MG
Fipronil sulfone is the sulfone metabolite of fipronil supplied as an analytical standard for research and method development. It is used as a reference material in studies of metabolism, environmental monitoring, and receptor-binding assays, and is reported to selectively inhibit the GABA receptor (IC50 ~175 nM).
  • High-purity analytical standard (reported 99.8%).
  • Molecular formula C12H4Cl2F6N4O2S and molecular weight 453.15 g/mol.
  • Suitable for method validation and quantification of fipronil metabolites.
  • Demonstrated GABA receptor activity useful for biochemical studies.
  • Available in small pack sizes appropriate for analytical workflows.