Learn More
Apexbio Technology LLC Firategrast 402567-16-2 5mg
SDP
Catalog No. 502240263
Encompass_Preferred
Firategrast (CAS 402567-16-2) is a small-molecule antagonist targeting integrins 4 1 and 4 7 It is designed to block integrin-ligand interactions thereby modulating leukocyte trafficking across the vascular endothelium into tissues Firategrast exerts its biological activity primarily through inhibition of leukocyte adhesion and migration particularly suppressing cell entry into the central nervous system (CNS) Based on these pharmacological properties Firategrast holds research potential in the treatment of relapsing-remitting multiple sclerosis (MS) by reducing CNS inflammation via inhibition of leukocyte infiltration
This item is not returnable. View return policy
Catalog No. 50-224-0263 Supplier Apexbio Technology LLC Supplier No. A34155
Firategrast (CAS 402567-16-2) is a small-molecule antagonist targeting integrins 4 1 and 4 7 It is designed to block integrin-ligand interactions thereby modulating leukocyte trafficking across the vascular endothelium into tissues Firategrast exerts its biological activity primarily through inhibition of leukocyte adhesion and migration particularly suppressing cell entry into the central nervous system (CNS) Based on these pharmacological properties Firategrast holds research potential in the treatment of relapsing-remitting multiple sclerosis (MS) by reducing CNS inflammation via inhibition of leukocyte infiltration