Learn More
Medchemexpress LLC HY-14951 5mg Medchemexpress, Firategrast CAS:402567-16-2 Purity:>98%
SDP
Catalog No. 502030570
Encompass_Preferred
Medchemexpress, HY-14951 5mg Firategrast CAS:402567-16-2 Firategrast is an orally bioavailable α4β1/α4β7 integrin antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0570 Supplier Medchemexpress LLC Supplier No. HY149515MG
Medchemexpress, HY-14951 5mg Firategrast CAS:402567-16-2 Firategrast is an orally bioavailable α4β1/α4β7 integrin antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.