Learn More
Medchemexpress LLC Firazorexton hydrate | 2861934-86-1 | 99.9% | 497.53 | C22H25F3N2O4S.3/2H2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY137440A5MG
Encompass_Preferred
Firazorexton hydrate is a research-grade small molecule that acts as a selective orexin type 2 receptor (OX2R) agonist. Supplied as a solid hydrate, it is orally active and brain-penetrant in preclinical studies and has been characterized for receptor signaling and wake-promoting activity in animal models, making it suitable for in vitro OX2R pharmacology and in vivo sleep/wake research.
  • Selective orexin type 2 receptor (OX2R) agonist.
  • Orally active and brain-penetrant in preclinical models.
  • Potent in vitro activity (EC50 ≈ 19 nM for calcium mobilization).
  • Demonstrated wake-promoting effects in mouse studies.
  • Supplied as a white to off-white solid hydrate with high purity.
Catalog No. 50-002-22807