Learn More
Medchemexpress LLC Firazorexton hydrate | 2861934-86-1 | 99.9% | C22H25F3N2O4S.3/2H2O | 10MG
SDP
Supplier:  Medchemexpress LLC HY137440A10MG
Encompass_Preferred
Firazorexton hydrate is a research-grade small molecule and selective orexin type 2 receptor (OX2R) agonist. It is orally active and brain-penetrant, and is used in preclinical pharmacology and in vivo studies of sleep-wake regulation such as narcolepsy models. The compound is supplied as a hydrate with high reported purity for laboratory research.
  • Selective orexin type 2 receptor (OX2R) agonist.
  • Orally active and brain-penetrant.
  • Used in preclinical pharmacology and in vivo sleep-wake studies.
  • Supplied as a hydrate with high reported purity.
  • Available in small research package sizes.
Catalog No. 50-002-59631