Learn More
Grainger Fire Bell 10 In.
Catalog No. 502766689
Encompass_Preferred
The Edwards Signaling Fire Bell is a 20 to 24 volt DC powered fire alarm bell with a loud, penetrating sound. It has a rugged steel housing finished in red enamel and operates on continuous voltage. The bell produces a sound output of 84 to 88 decibels at 10 feet. It is weatherproof and suitable for indoor or outdoor use. The bell measures 5 inches high by 5 inches wide.
Catalog No. 50-276-6689 Supplier Grainger Supplier No. 16X290
The Edwards Signaling Fire Bell is a 20 to 24 volt DC powered fire alarm bell with a loud, penetrating sound. It has a rugged steel housing finished in red enamel and operates on continuous voltage. The bell produces a sound output of 84 to 88 decibels at 10 feet. It is weatherproof and suitable for indoor or outdoor use. The bell measures 5 inches high by 5 inches wide.