Learn More
Grainger Fire Bell 8 In.
Catalog No. 502767541
Encompass_Preferred
The Edwards Signaling Fire Bell is a 6 inch diameter, red painted steel fire alarm bell. It produces a loud, penetrating sound to alert building occupants of a fire or other emergency. The bell operates on 24 volts DC and draws 0.33 amps. It is weatherproof and suitable for indoor or outdoor use. The bell has an adjustable volume from 80 to 102 decibels. It meets NFPA 72 standards for audible signaling appliances. The bell comes with a mounting bracket for wall or ceiling mounting. It is made in the USA.
Catalog No. 50-276-7541 Supplier Grainger Supplier No. 16X292
The Edwards Signaling Fire Bell is a 6 inch diameter, red painted steel fire alarm bell. It produces a loud, penetrating sound to alert building occupants of a fire or other emergency. The bell operates on 24 volts DC and draws 0.33 amps. It is weatherproof and suitable for indoor or outdoor use. The bell has an adjustable volume from 80 to 102 decibels. It meets NFPA 72 standards for audible signaling appliances. The bell comes with a mounting bracket for wall or ceiling mounting. It is made in the USA.