Learn More
Grainger Fire Bell Gray 10 In.
Catalog No. 502767553
Encompass_Preferred
The Edwards Signaling 24V DC Fire Bell is a signaling device designed for fire alarm systems. It produces a loud, penetrating sound to alert building occupants of a fire emergency. This bell operates on 24V DC and draws 0.18 amps. It features an ABS plastic housing with a red finish and produces a sound output of 84 dB at 10 feet. The bell is weatherproof and suitable for indoor or outdoor use. It can be flush mounted on walls or ceilings. The bell is UL Listed and meets NFPA 72 requirements. With dimensions of 5.5" diameter x 2.75" depth, this fire bell provides an audible signal for fire alarm notification.
Catalog No. 50-276-7553 Supplier Grainger Supplier No. 16X279
The Edwards Signaling 24V DC Fire Bell is a signaling device designed for fire alarm systems. It produces a loud, penetrating sound to alert building occupants of a fire emergency. This bell operates on 24V DC and draws 0.18 amps. It features an ABS plastic housing with a red finish and produces a sound output of 84 dB at 10 feet. The bell is weatherproof and suitable for indoor or outdoor use. It can be flush mounted on walls or ceilings. The bell is UL Listed and meets NFPA 72 requirements. With dimensions of 5.5" diameter x 2.75" depth, this fire bell provides an audible signal for fire alarm notification.