Learn More
Grainger Edwards Signaling Fire Bell, 10 In., 20 To 24 V DC, Gray, 1 Ea.
Catalog No. 502767551
Encompass_Preferred
These underdome fire alarm bells are engineered for reliable performance in fire alarm systems. Constructed with a 10 in. selected alloy steel gong and a heavy-duty mechanism, they deliver a loud, resonant tone designed to overcome ambient noise. The unit is diode polarized and features a rugged, compact design suitable for surface mounting on walls or ceilings.
  • Operating voltage of 20 V DC to 24 V DC
  • Peak output of 86 dBA
  • Heavy-duty cast housing with gray finish and red label
  • Suitable for indoor or weatherproof applications when paired with optional back box
  • Fits any standard single-gang opening
  • Low power drain and wide voltage selection
  • FM and UL approved
Catalog No. 50-276-7551 Supplier Grainger Supplier No. 16X285
These underdome fire alarm bells are engineered for reliable performance in fire alarm systems. Constructed with a 10 in. selected alloy steel gong and a heavy-duty mechanism, they deliver a loud, resonant tone designed to overcome ambient noise. The unit is diode polarized and features a rugged, compact design suitable for surface mounting on walls or ceilings.
  • Operating voltage of 20 V DC to 24 V DC
  • Peak output of 86 dBA
  • Heavy-duty cast housing with gray finish and red label
  • Suitable for indoor or weatherproof applications when paired with optional back box
  • Fits any standard single-gang opening
  • Low power drain and wide voltage selection
  • FM and UL approved