Learn More
Grainger Fire Bell Gray 6 in 20 to 24V
Catalog No. 502767552
Encompass_Preferred
The Edwards Signaling 24V DC Fire Bell is a 6 inch diameter bell made of red painted steel with a gong tone. It produces a loud, penetrating sound and is designed for fire alarm applications. This bell operates on 24V DC and draws 0.18 amps. It has an adjustable volume between 80 to 102 decibels. The bell is weatherproof and includes a mounting bracket for wall or ceiling mounting. It is UL Listed and meets the requirements of NFPA 72. The overall dimensions are 6.5 inches high by 6 inches diameter. This is an alarm signaling device well suited for fire detection systems.
Catalog No. 50-276-7552 Supplier Grainger Supplier No. 16X287
The Edwards Signaling 24V DC Fire Bell is a 6 inch diameter bell made of red painted steel with a gong tone. It produces a loud, penetrating sound and is designed for fire alarm applications. This bell operates on 24V DC and draws 0.18 amps. It has an adjustable volume between 80 to 102 decibels. The bell is weatherproof and includes a mounting bracket for wall or ceiling mounting. It is UL Listed and meets the requirements of NFPA 72. The overall dimensions are 6.5 inches high by 6 inches diameter. This is an alarm signaling device well suited for fire detection systems.