Learn More
Grainger Fire Bell Gray 6 IN.
Catalog No. 502767177
Encompass_Preferred
The Edwards Signaling Fire Bell Vibrating is a 6 inch diameter vibrating fire alarm bell made of steel with a red powder coated finish. It produces a loud, pulsating warning sound at 88 decibels that can be heard even in noisy environments. The bell requires 24 VDC and draws 0.33 amps. It is weatherproof and suitable for indoor or outdoor use. The vibrating bell alarm is UL listed and meets NFPA 72 requirements.
Catalog No. 50-276-7177 Supplier Grainger Supplier No. 16X281
The Edwards Signaling Fire Bell Vibrating is a 6 inch diameter vibrating fire alarm bell made of steel with a red powder coated finish. It produces a loud, pulsating warning sound at 88 decibels that can be heard even in noisy environments. The bell requires 24 VDC and draws 0.33 amps. It is weatherproof and suitable for indoor or outdoor use. The vibrating bell alarm is UL listed and meets NFPA 72 requirements.