Learn More
Grainger Edwards Signaling Vibrating Bell, Alloy Steel, 120V AC, Red, 8 in.
Catalog No. 502766694
Encompass_Preferred
These diode polarized, vibrating fire alarm bells are designed for reliable integration with fire alarm control equipment. Featuring a rugged construction, these bells are suitable for audible signaling in diverse installations, operating in conjunction with an installed fire alarm panel and detection devices.
  • Resonant tone gong made of alloy steel
  • Epoxy powder coated finish in red
  • Designed for surface mounting
  • Low power drain
  • Operates at 120 V AC (60 Hz)
  • 86 dBA output at 10 ft. (3.05 m)
  • Suitable for ambient temperatures from -31°F to +150°F
  • UL listed and FM approved
Catalog No. 50-276-6694 Supplier Grainger Supplier No. 16X284
These diode polarized, vibrating fire alarm bells are designed for reliable integration with fire alarm control equipment. Featuring a rugged construction, these bells are suitable for audible signaling in diverse installations, operating in conjunction with an installed fire alarm panel and detection devices.
  • Resonant tone gong made of alloy steel
  • Epoxy powder coated finish in red
  • Designed for surface mounting
  • Low power drain
  • Operates at 120 V AC (60 Hz)
  • 86 dBA output at 10 ft. (3.05 m)
  • Suitable for ambient temperatures from -31°F to +150°F
  • UL listed and FM approved