Learn More
Grainger Bullard Fire Helmet, Thermoplastic, Black, 6 1/2 to 8 in., 1 ea
Catalog No. 502771999
Encompass_Preferred
This black fire helmet is engineered for structural and proximity firefighting, featuring a high-heat thermoplastic shell for superior impact and degradation resistance. Designed with a low center of gravity for balance and stability, it incorporates a 36-point comfort system to provide a custom-fitted experience for users.
  • Exceeds NFPA 1971 standards for structural and proximity fire fighting.
  • High-heat thermoplastic outer shell provides greater impact and penetration resistance than fiberglass.
  • Adjustable ride and balance with 36 custom comfort settings.
  • Reflective striping retains reflectivity up to 500°F (260°C).
  • Molded inner liner impact cap system provides thermal rise protection and shell security.
  • Integrated goggle grommets enhance comfort and support.
  • Includes 4 in. faceshield and integrated lighting system.
Catalog No. 50-277-1999 Supplier Grainger Supplier No. 13W789
This black fire helmet is engineered for structural and proximity firefighting, featuring a high-heat thermoplastic shell for superior impact and degradation resistance. Designed with a low center of gravity for balance and stability, it incorporates a 36-point comfort system to provide a custom-fitted experience for users.
  • Exceeds NFPA 1971 standards for structural and proximity fire fighting.
  • High-heat thermoplastic outer shell provides greater impact and penetration resistance than fiberglass.
  • Adjustable ride and balance with 36 custom comfort settings.
  • Reflective striping retains reflectivity up to 500°F (260°C).
  • Molded inner liner impact cap system provides thermal rise protection and shell security.
  • Integrated goggle grommets enhance comfort and support.
  • Includes 4 in. faceshield and integrated lighting system.