Learn More
Grainger BULLARD Fire Helmet, Thermoplastic, Lime Yellow, 6 1/2 in. to 8 in. 1 ea
Catalog No. 502773615
Encompass_Preferred
This modern fire helmet is designed for durability and protection in high-risk environments. Constructed from high-performance thermoplastic, the unit features an integrated suspension and faceshield system for optimal safety and comfort.
  • Features a 4-point ratchet suspension system
  • Includes a 10.2 cm (4 in.) faceshield
  • Equipped with a quick release and postman slide chin strap
  • Fits head sizes from 6 1/2 in. to 8 in.
  • Constructed from thermoplastic material
  • Color: lime yellow
Catalog No. 50-277-3615 Supplier Grainger Supplier No. 13W807
This modern fire helmet is designed for durability and protection in high-risk environments. Constructed from high-performance thermoplastic, the unit features an integrated suspension and faceshield system for optimal safety and comfort.
  • Features a 4-point ratchet suspension system
  • Includes a 10.2 cm (4 in.) faceshield
  • Equipped with a quick release and postman slide chin strap
  • Fits head sizes from 6 1/2 in. to 8 in.
  • Constructed from thermoplastic material
  • Color: lime yellow