Learn More
Grainger Bullard LTX Structural Fire Helmet, Thermoplastic, Orange, 1 ea
Catalog No. 502771916
Encompass_Preferred
The LTX structural fire helmet features a modern, flared, rear-brim design engineered to help protect firefighters from head and neck injuries. Constructed from heat-resistant thermoplastic, this helmet incorporates an integrated impact management system consisting of an outer shell, a unitary inner shell, and a crown strap suspension to reduce impact forces during structural firefighting.
  • Meets NFPA 1971 standards for protective ensembles.
  • Features a heat-resistant thermoplastic shell with aluminum-reinforced, elastomeric edge beading.
  • Includes a 4-point ratchet headband for quick-adjustment sizing, fitting hat sizes 6-1/2 in. to 8 in.
  • Equipped with a removable, cushion-backed leather ratchet cover for comfort.
  • Includes a 4 in. faceshield.
  • Provides a 5-year warranty on the shell and a 10-year lifetime warranty on non-electronic components.
Catalog No. 50-277-1916 Supplier Grainger Supplier No. 13W806
The LTX structural fire helmet features a modern, flared, rear-brim design engineered to help protect firefighters from head and neck injuries. Constructed from heat-resistant thermoplastic, this helmet incorporates an integrated impact management system consisting of an outer shell, a unitary inner shell, and a crown strap suspension to reduce impact forces during structural firefighting.
  • Meets NFPA 1971 standards for protective ensembles.
  • Features a heat-resistant thermoplastic shell with aluminum-reinforced, elastomeric edge beading.
  • Includes a 4-point ratchet headband for quick-adjustment sizing, fitting hat sizes 6-1/2 in. to 8 in.
  • Equipped with a removable, cushion-backed leather ratchet cover for comfort.
  • Includes a 4 in. faceshield.
  • Provides a 5-year warranty on the shell and a 10-year lifetime warranty on non-electronic components.