Learn More
Sigma Aldrich Fine Chemicals Biosciences fire Monkey fire flower
Supplier:  Sigma Aldrich Fine Chemicals Biosciences REV888SAMPLE
Encompass
for the extraction of high molecular weight DNA from animal and bacterial cells and size selection of extracted DNA from all sample types.fIRE MONKEY - HMW DNA EXTRACTIONfire Monkey is a standard spin-column process which extracts HMW-DNA with average strand lengths of 100kb and above from both bacterial and mammalian cells within 1hr. fIRE fLOWER - DNA SIZE SELECTION KITfire flower is a standard spin-column process that can size select extracted DNA from all sample sources within 15mins. fire flower can increase the overall average strand length by 30% while depleting DNA fragments up to 30kb. fire flower can be used to size-select up to 200 muL of extracted DNA despite the sample being rich in small DNA fragments.The two major disruptive features of fire Monkey are that firstly it generates average DNA fragment lengths of between 100kbp - 130kbp and secondly it significantly reduces the amount of small DNA fragments shorter than 10kbp in the final product. This combination where
Catalog No. 50-290-6562