Learn More
Grainger Fire Panel Trim 10 Zone Red
Catalog No. 502766203
Encompass_Preferred
The Edwards Signaling 10-Zone Fire Panel Trim is a fire alarm control panel accessory that expands the zone capacity of compatible Edwards fire alarm control panels by 10 zones. It features screw terminal connections and mounts inside the control panel cabinet. This trim is compatible with the Edwards EST3 Series, EST2 Series, and EST fire alarm control panels when used with the proper personality module. It requires a 24VDC power connection from the host control panel. The dimensions are 10.5" x 5.25" x 1.25". This 10-zone expansion module is UL Listed and made in the USA by Edwards Signaling.
Catalog No. 50-276-6203 Supplier Grainger Supplier No. 16X407
The Edwards Signaling 10-Zone Fire Panel Trim is a fire alarm control panel accessory that expands the zone capacity of compatible Edwards fire alarm control panels by 10 zones. It features screw terminal connections and mounts inside the control panel cabinet. This trim is compatible with the Edwards EST3 Series, EST2 Series, and EST fire alarm control panels when used with the proper personality module. It requires a 24VDC power connection from the host control panel. The dimensions are 10.5" x 5.25" x 1.25". This 10-zone expansion module is UL Listed and made in the USA by Edwards Signaling.