Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Fire Protection Water High Performance Pipe Marker, White on Red

Catalog No. 18038112 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-038-112 4 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-038-112 Supplier Brady™ Supplier No. 5689OO
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wrap Around High Performance Pipe Markers (B-689) are coiled, printed plastic sheets with sealed color graphics for use on pipes in harsh environments.

  • Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive labels aren’t effective.
  • Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
  • Self-adhesive end strip adheres to the marker's body to ensure a tight, continuous wrap.
  • Excellent chemical resistance.

Specifications

Adhesive Type Non-adhesive
Coating Laminate
Color White on Red
Display Text
Finish Gloss
Format Sheet
Material Coiled Plastic (B-689)
Product Type Pipe Marker
Sterility Non-sterile
Dimensions (L x W) 4 x 4 in.
Length (English) 4 in.
Width (English) 4 in.
Width (Metric) 101.60 mm
Diameter (English) 0.375 to 0.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.