Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Firefly In A Flask: Oscillating Chemiluminescence
SCI_ED

Catalog No. S01982 Shop All Aldon Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01982 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01982 Supplier Innovating Science™ Supplier No. IS7052
Add to Cart
Add to Cart

Two glow-in-the-dark reactions occur in turn: one that is bright an quick, and another that is dull and slow. The oscillation between these reactions looks like a firefly on a summer night!

Two glow-in-the-dark reactions occur in turn: one that is bright an quick, and another that is dull and slow. The oscillation between these reactions looks like a firefly on a summer night!

Specifications

Grade 6+
Class Size 5 Demonstrations
Product Type Spectrophotometric Analysis of Copper, Beer's Law Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.