Learn More
Sigma Aldrich Fine Chemicals Biosciences firefly Luciferase Assay f
Catalog No. 502904555
Encompass
firefly luciferase is widely used as a reporter for studying gene regulation and function and for pharmaceutical screening. It is a very sensitive genetic reporter due to the absence of endogenous luciferase activity in mammalian cells or tissues. firefly luciferase is a 62 000 Dalton protein which is active as a monomer and does not require subsequent processing for its activity. The enzyme catalyzes ATP-dependent D-luciferin oxidation to oxyluciferin producing light emission centered at 560 nm. Light emitted from the reaction is directly proportional to the number of luciferase enzyme molecules. The firefly Luciferase Assay is designed for simple and efficient quantitation of firefly luciferase reporter enzyme activity from cultured cells with high sensitivity and linearity. This is a flash-type luminescence assay that requires signal to be measured immediately after adding working SOLUTIONon to samples. The luminescence signal decays over the course of about 10 minutes of reaction
Catalog No. 50-290-4555 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCT154
firefly luciferase is widely used as a reporter for studying gene regulation and function and for pharmaceutical screening. It is a very sensitive genetic reporter due to the absence of endogenous luciferase activity in mammalian cells or tissues. firefly luciferase is a 62 000 Dalton protein which is active as a monomer and does not require subsequent processing for its activity. The enzyme catalyzes ATP-dependent D-luciferin oxidation to oxyluciferin producing light emission centered at 560 nm. Light emitted from the reaction is directly proportional to the number of luciferase enzyme molecules. The firefly Luciferase Assay is designed for simple and efficient quantitation of firefly luciferase reporter enzyme activity from cultured cells with high sensitivity and linearity. This is a flash-type luminescence assay that requires signal to be measured immediately after adding working SOLUTIONon to samples. The luminescence signal decays over the course of about 10 minutes of reaction