Learn More
Biotium Firefly Luciferase Assay Kit 2.0, 1000 Assays
Catalog No. NC1254223
Encompass_Preferred
Firefly Luciferase Assay Kit 2.0 is a flash-type assay for firefly luciferase activity in transfected cell lysates. This kit is an improved version of our original Firefly Luciferase Assay (catalog no. 30003), with better working solution stability. 1000 assays.
Catalog No. NC1254223 Supplier Biotium Supplier No. 300852
Firefly Luciferase Assay Kit 2.0 is a flash-type assay for firefly luciferase activity in transfected cell lysates. This kit is an improved version of our original Firefly Luciferase Assay (catalog no. 30003), with better working solution stability. 1000 assays.