Learn More
BPS Bioscience Inc Firefly Luciferase - CHO Recombinant Cell Line , 2 Vials
SDP
Catalog No. NC3243005
Encompass
Recombinant CHO-K1 cells constitutively expressing the firefly luciferase reporter.This product has been cited 6 times.Purchase of this cell line is for research purposes only; commercial use requires a separate license. View the full terms and conditions.
Catalog No. NC3243005 Supplier BPS Bioscience Inc Supplier No. 79725
Recombinant CHO-K1 cells constitutively expressing the firefly luciferase reporter.This product has been cited 6 times.Purchase of this cell line is for research purposes only; commercial use requires a separate license. View the full terms and conditions.