Learn More
BPS Bioscience Inc Firefly Luciferase-eGFP Lentivirus, 2 vials of 500 μl, HIV-based, VSV-G pseudotyped, Replication incompetent, EF1A promoter, Geneticin (G418) selection
SDP
Catalog No. NC3622041
Encompass
Firefly Luciferase-eGFP Lentivirus, 2 vials of 500 μl, HIV-based, VSV-G pseudotyped, Replication incompetent, EF1A promoter, Geneticin (G418) selection
Catalog No. NC3622041 Supplier BPS Bioscience Inc Supplier No. 78741G
Firefly Luciferase-eGFP Lentivirus, 2 vials of 500 μl, HIV-based, VSV-G pseudotyped, Replication incompetent, EF1A promoter, Geneticin (G418) selection